Learn More
Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Zinc nitrate hexahydrate is used as an analytical reagent, catalyst and as a mordant in dyeing. It finds application in electro galvanizing, rubber industry and in the water treatment industry. It is widely used as the precursor for the synthesis of zinc oxide. It is employed in the preparation of the steel phosphating agent. In the field of agriculture, it is used for nourishing and protecting fruit trees from acid rain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼36°C |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Linear Formula | Zn(NO3)2·6H2O |
UN Number | UN1514 |
Merck Index | 14,10144 |
Solubility Information | Soluble in water and alcohol. |
IUPAC Name | zinc;dinitrate;hexahydrate |
Formula Weight | 297.47 (189.38 Anhydrous) |
Show More |
RUO – Research Use Only