missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™

£86.81 - £113.16

Chemical Identifiers

CAS 10196-18-6
Molecular Formula C6H26N2O6Zn
Molecular Weight (g/mol) 287.66
MDL Number MFCD00149889
InChI Key PZDYRUXUUCEJGL-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
IUPAC Name zinc;dinitrate;hexahydrate
SMILES C.C.C.C.C.C.[Zn].O[N+]([O-])=O.O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10796301
SDS View Documents
Fisher Chemical
Z/1150/50
250g £86.81
250g
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
10020360
SDS View Documents
Fisher Chemical
Z/1150/53
500g £113.16
500g
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

10196-18-6
287.66
PZDYRUXUUCEJGL-UHFFFAOYSA-N
15865313
C.C.C.C.C.C.[Zn].O[N+]([O-])=O.O[N+]([O-])=O
C6H26N2O6Zn
MFCD00149889
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
zinc;dinitrate;hexahydrate

Specifications

10196-18-6
MFCD00149889
PZDYRUXUUCEJGL-UHFFFAOYSA-N
zinc;dinitrate;hexahydrate
15865313
Solid
White
5.1
C6H26N2O6Zn
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
C.C.C.C.C.C.[Zn].O[N+]([O-])=O.O[N+]([O-])=O
287.66
297.46g/mol
Plastic powder jar
36°C
250g
Safety and Handling

Safety and Handling

Product Identifier
  • Zinc nitrate hexahydrate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Oxidising solid Category 2

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS