missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc nitrate hexahydrate, 99% (metals basis), Thermo Scientific Chemicals
30.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
10 kg
Description
Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∽36°C |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in water,alcoholSoluble in water and ethanol. |
Formula Weight | 297.47 (189.38 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 2.07 g/mL |
Chemical Name or Material | Zinc nitrate hexahydrate |
Safety and Handling
Product Identifier
- Zinc nitrate hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Nitric acid, zinc salt, hexahydrate, zinc nitrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'