missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number p-4509951
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
10196-18-6
H12N2O12Zn
297.48
MFCD00149889
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
15865313
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
10196-18-6
H12N2O12Zn
297.48
MFCD00149889
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
15865313
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 36.0°C
Color White
Physical Form Adhering Crystals
Quantity 250 g
Assay Percent Range 98%
Linear Formula Zn(NO3)2·6H2O
Merck Index 14,10144
Solubility Information Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol
IUPAC Name zinc(2+) hexahydrate dinitrate
Formula Weight 297.46
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Zinc nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'