missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 98%, extra pure, ACROS Organics™

Brand:  Acros Organics 211660051

Code : 29

Additional Details : CAS Number : 10196-18-6 Weight : 5.05000kg Transport : UN number : 1514 Chem class : 1 Pack group : II

Quantity 5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 11944271

  • £210.30 / 5kg


Estimated Shipment
08-02-2018
Add to basket

Specifications

Chemical Identifiers

Zinc nitrate hexahydrate
Extra Pure
98%
Zn(NO3)2·6H2O
10196-18-6
98%
N2O6Zn·6H2O
297.46

Specifications

Zinc nitrate hexahydrate
Extra Pure
98%
N2O6Zn·6H2O
297.46
20ppm max.
Plastic bottle
White
5kg
10196-18-6
Adhering Crystals
98%
Zn(NO3)2·6H2O
100ppm max.
100ppm max.
Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol
36.0°C
Videos
SDS