missing translation for 'onlineSavingsMsg'
Learn More

Zinc iron oxide, Thermo Scientific Chemicals

Catalog Number p-7049723
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
12063-19-3
Fe2H2O4Zn
243.082
MFCD00049621
NNGHIEIYUJKFQS-UHFFFAOYSA-L
Zinc ferrite
11831558
hydroxy(oxo)iron;zinc
O[Fe]=O.O[Fe]=O.[Zn]
This item is not returnable. View return policy
12063-19-3
Fe2H2O4Zn
243.082
MFCD00049621
NNGHIEIYUJKFQS-UHFFFAOYSA-L
Zinc ferrite
11831558
hydroxy(oxo)iron;zinc
O[Fe]=O.O[Fe]=O.[Zn]

Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Fine Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 241.06
Odor Odorless
Chemical Name or Material Zinc iron oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'