missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, 98%, Thermo Scientific Chemicals

Catalog Number p-7048853
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
10139-47-6
MFCD00011299
This item is not returnable. View return policy
10139-47-6
MFCD00011299

Zinc iodide is often used as an x-ray opaque penetrant in industrial radiography to improve the contrast between the damage and intact composite.n conjunction with osmium tetroxide ZnI2 is used as a stain in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 625°C (decomposition)
Melting Point 446°C
Color White to Yellow
Physical Form Crystalline
Quantity 50 g
Linear Formula ZnI2
UN Number UN3260
Merck Index 14,10140
Solubility Information Freely soluble in water, glycerol, alcohol, ether
Formula Weight 319.18
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 4.74 g/mL
Chemical Name or Material Zinc iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'