missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc iodide, 98%, Thermo Scientific Chemicals
37.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Zinc iodide is often used as an x-ray opaque penetrant in industrial radiography to improve the contrast between the damage and intact composite.n conjunction with osmium tetroxide ZnI2 is used as a stain in electron microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 625°C (decomposition) |
Melting Point | 446°C |
Color | White to Yellow |
Physical Form | Crystalline |
Quantity | 50 g |
Linear Formula | ZnI2 |
UN Number | UN3260 |
Merck Index | 14,10140 |
Solubility Information | Freely soluble in water, glycerol, alcohol, ether |
Formula Weight | 319.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'