missing translation for 'onlineSavingsMsg'
Learn More

Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals

Catalog Number 11308048
Click to view available options
Quantity:
100 g
500 g
1 kg
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]

Zinc Granules; ACS; 99.8%; Alfa Aesar; -20 Mesh

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.8%
Quantity 500 g
Grade ACS Reagent
Mesh Size -20 Mesh
Form Granular
Chemical Name or Material Zinc Granules, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'