missing translation for 'onlineSavingsMsg'
Learn More

Zinc formate dihydrate, 98%, Alfa Aesar™

Brand:  Alfa Aesar 013797.A1

Code : NEW

Additional Details : CAS Number : 5970-62-7 Weight : 1.00000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 1kg

 View more versions of this product

Product Code. 11311339

  • £65.93 / 1kg


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

Zinc formate dihydrate
Powder
Zn(CHO2)2·2H2O
MFCD00050836
5970-62-7
98%
191.43 (155.40 Anhydrous)

Specifications

Zinc formate dihydrate
Powder
Zn(CHO2)2·2H2O
MFCD00050836
5970-62-7
98%
191.43 (155.40 Anhydrous)
1kg
Videos
SDS