missing translation for 'onlineSavingsMsg'
Learn More

Zinc Fluoride, 99%, Honeywell™ Fluka™

Brand:  Honeywell Fluka 205567-100G

Code : NEW

Additional Details : CAS Number : 7783-49-5 Weight : 0.10000kg

Packaging Poly Bottle
Quantity 100g

 View more versions of this product

Product Code. 15641770

  • £67.23 / 100g


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

1mmHg (970°C)
ZnF2
99%
Powder
7783-49-5
White
4.95g/mL at 25°C
MFCD00011298
103.39g/mol
Zinc Fluoride
872°C

Specifications

Zinc Fluoride
Powder
ZnF2
MFCD00011298
Poly Bottle
White
100g
7783-49-5
99%
103.39g/mol
4.95g/mL at 25°C
1mmHg (970°C)
872°C
Videos