missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is used for a source of the zinc ion. It can also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. It is used as a moderate-strength lewis acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Linear Formula | ZnCl2·xH2O |
UN Number | UN3260 |
Merck Index | 14,10132 |
Solubility Information | Soluble in water,ethanol (slightly),acetone (slightly),ether,and glycerol. |
SMILES | Cl[Zn]Cl |
IUPAC Name | dichlorozinc |
Formula Weight | 136.29 (Anhydrous) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'