missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, Free-flowing, Reagent Grade, anhydrous, ≥98%, Honeywell Fluka™

Catalog Number p-7111984
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Free-flowing, Reagent Grade, anhydrous,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 293°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 500 g
Linear Formula ZnCl2
UN Number UN2331
Formula Weight 136.3
Percent Purity >97%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.93 g/cm3
Chemical Name or Material Zinc chloride, Free-flowing, Reagent Grade, anhydrous, ≥98%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'