missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Boiling Point | 732°C |
Melting Point | 293°C |
Color | White |
pH | 5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 136.29g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1.3mbar 428 |
Safety and Handling
Product Identifier
- Zinc chloride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'