missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, ≥97%, Honeywell™ Fluka™

Brand:  Honeywell Fluka 746355-1KG

Code : NEW

Additional Details : CAS Number : 7646-85-7 Weight : 1.00000kg Transport : UN number : 2331 Chem class : 8 Pack group : III

Packaging Poly bottle
Quantity 1kg

 View more versions of this product

Product Code. 15636960

  • £84.33 / 1kg


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

≥97%
Zinc chloride
MFCD00011295
136.3g/mol
7646-85-7
UN2331

Specifications

Zinc chloride
≥97%
136.3g/mol
UN2331
Odorless
851g/L
Solid
287° to 304°C
1kg
7646-85-7
ZnCl2
MFCD00011295
2.93g/cm3
Poly bottle
1mmHg (428°C)
White
5 at 20°C
Videos
SDS