missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, 1M in diethyl ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
32.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | ZnCl2 |
UN Number | UN2924 |
Flash Point | -40°C (-40°F) |
Merck Index | 14,10132 |
Formula Weight | 136.29 |
Concentration or Composition (by Analyte or Components) | 1M in diethyl ether |
Packaging | ChemSeal™ bottles |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'