missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 1M in diethyl ether, Thermo Scientific Chemicals

Catalog Number 15431087
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 500 mL
Linear Formula ZnCl2
UN Number UN2924
Flash Point -40°C (-40°F)
Merck Index 14,10132
Formula Weight 136.29
Concentration or Composition (by Analyte or Components) 1M in diethyl ether
Sensitivity Hygroscopic
Density 0.835 g/mL
Chemical Name or Material Zinc chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'