missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Certified AR for Analysis, Granulated Metal, Fisher Chemical™

Brand:  Fisher Chemical Z/0300/60

20.00 GBP valid until 2021-10-31
Use promo code "16370" to get your promotional price.



Code : 22

Additional Details : CAS Number : 7440-66-6 Weight : 1.00000kg

 View more versions of this product

Product Code. 10182400

  • £149.00 / 1kg

In stock, ready to ship: 9

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zinc, Certified AR for Analysis, Granulated Metal

Specifications

Specifications

1kg
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
23994
65.36g/mol
1.3mbar 478
419°C
7440-66-6
11291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
Plastic powder jar
CHEBI:30185
Solid Various Form
Gray
907°C
Videos
SDS Documents
Certificates
View accessory products