missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059232
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
299465-28-4
Br2Zn
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
This item is not returnable. View return policy
299465-28-4
Br2Zn
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208

Zinc bromide is an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. Zinc bromide is employed as a lewis acid in organic chemistry. Has been used as a transparent shield against radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 100 g
UN Number UN3260
Merck Index 14,10129
Solubility Information Soluble in water (4470mg/ml at 20°C),ethanol,ether,90% alcohol (2000mg/ml),and water (4710mg/ml at 25°C).
SMILES [Zn++].[Br-].[Br-]
IUPAC Name zinc(2+) dibromide
Molecular Weight (g/mol) 225.19
Formula Weight 225.19 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'