missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11319738
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.188
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.188
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]

Mainly used in ocean wells as complete liquid, stationary liquid, amending liquid. It is also an important material in medical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Boiling Point 650°C
Melting Point 394°C
Color White
Physical Form Powder
Quantity 50 g
UN Number UN3260
Merck Index 14,10129
Solubility Information Very soluble in ethanol and soluble in ethyl ether. Soluble in water (at 20°C 4470g/L).
Formula Weight 225.19
Percent Purity 99.999%
Refractive Index 1.5452
Sensitivity Hygroscopic
Density 4.201 g/mL
Assay (metals basis)
Chemical Name or Material Zinc bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'