missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog Number 10484335
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.19
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.19
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Boiling Point 697.0°C
Melting Point 394.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.999%
Linear Formula ZnBr2
Fieser 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389
Merck Index 15, 10328
Solubility Information Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether,solutions of alkali hydroxides,1g/0.5mL 90% alcohol
Formula Weight 225.19
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.22
Density 4.2200g/mL
Chemical Name or Material Zinc bromide, 99.999%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'