missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc bromide, 99.9% (metals basis), Thermo Scientific Chemicals
32.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
In organic chemistry as a Lewis acid. It is the electrolyte in the zinc bromide battery. Zinc bromide solutions can be used as a transparent shield against radiation. In oil and natural gas wells, solutions containing zinc bromide are used to displace drilling mud.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 650°C |
Melting Point | 394°C |
Color | Cream to White |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3260 |
Merck Index | 14,10129 |
Solubility Information | Very soluble in water,alcohol,ether,acetone,tetrahydrofuran. |
Formula Weight | 225.19 |
Percent Purity | 99.9% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'