missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 143°C to 147°C |
Color | White |
Flash Point | >110°C (230°F) |
Linear Formula | [(CF3SO2)2N]2Zn |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | It is soluble in CH2Cl2 and H2O. |
Formula Weight | 625.65 |
Physical Form | Solid |
Chemical Name or Material | Zinc bis (trifluoromethylsulfonyl)imide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'