missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, Certified AR for Analysis, Fisher Chemical™

£65.90 - £215.00

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number 66961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10795151
SDS View Documents
Fisher Chemical
Z/0700/50
250g £65.90
250g
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 32
Please to purchase this item. Need a web account? Register with us today!
10326060
SDS View Documents
Fisher Chemical
Z/0700/53
500g £115.00
500g
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 39
Please to purchase this item. Need a web account? Register with us today!
10417660
SDS View Documents
Fisher Chemical
Z/0700/60
1kg £215.00
1kg
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 7
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5970-45-6
219.498
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
C4H10O6Zn
66961
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
zinc;diacetate;dihydrate

Specifications

5970-45-6
66961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc;diacetate;dihydrate
2724192
Solid
White
6.0 to 7.0
C4H10O6Zn
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
219.498
219.5g/mol
Plastic powder jar
237°C
250g
Safety and Handling

Safety and Handling

Product Identifier
  • Zinc acetate dihydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Product Certifications