missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, Thermo Scientific Chemicals
96.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
Description
Zinc 2-methoxyethoxide is used as catalyst component in oxirane polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Density | 0.96 g/mL |
Boiling Point | 124°C |
Flash Point | 40°C (104°F) |
Linear Formula | Zn(OCH2CH2OCH3)2 |
Quantity | 25 mL |
UN Number | UN1188 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in methoxyethanol and pyridine. Miscible with alcohols,ketones,esters and water. |
Formula Weight | 215.56 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'