missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, Thermo Scientific Chemicals

Catalog Number 11324948
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
129918-15-6
C6H16O4Zn
217.57
MFCD00156529
SZCCOQAZCQRATH-UHFFFAOYSA-N
This item is not returnable. View return policy
129918-15-6
C6H16O4Zn
217.57
MFCD00156529
SZCCOQAZCQRATH-UHFFFAOYSA-N

Zinc 2-methoxyethoxide is used as catalyst component in oxirane polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula Zn(OCH2CH2OCH3)2
Quantity 100 mL
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Soluble in methoxyethanol and pyridine. Miscible with alcohols,ketones,esters and water.
Formula Weight 215.56
Physical Form Liquid
Chemical Name or Material Zinc 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'