missing translation for 'onlineSavingsMsg'
Learn More

Zeolite, type: ZSM-5, Thermo Scientific Chemicals

Catalog Number 10762101
Click to view available options
Quantity:
100 g
500 g
5 kg
This item is not returnable. View return policy
308081-08-5
C14H16N2
212.296
MFCD00132601
HSWPZIDYAHLZDD-UHFFFAOYSA-N
atipamezole, antisedan, 4-2-ethyl-2-indanyl imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, atipamezol spanish, atipamezolum latin, unii-03n9u5jaf6, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, dsstox_cid_29061
71310
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3
This item is not returnable. View return policy
308081-08-5
C14H16N2
212.296
MFCD00132601
HSWPZIDYAHLZDD-UHFFFAOYSA-N
atipamezole, antisedan, 4-2-ethyl-2-indanyl imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, atipamezol spanish, atipamezolum latin, unii-03n9u5jaf6, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, dsstox_cid_29061
71310
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Solubility in water: insoluble
Loss on Ignition 10 % max. (1000°C)
Packaging Plastic bottle
Chemical Name or Material Zeolite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'