missing translation for 'onlineSavingsMsg'
Learn More

Zeolite mordenite, ammonium, Thermo Scientific Chemicals

Catalog Number 15412517
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1318-02-1
Al2O5Si
162.043
MFCD00132601
HNPSIPDUKPIQMN-UHFFFAOYSA-N
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
9942228
dioxosilane;oxo(oxoalumanyloxy)alumane
O=[Al]O[Al]=O.O=[Si]=O
This item is not returnable. View return policy
1318-02-1
Al2O5Si
162.043
MFCD00132601
HNPSIPDUKPIQMN-UHFFFAOYSA-N
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
9942228
dioxosilane;oxo(oxoalumanyloxy)alumane
O=[Al]O[Al]=O.O=[Si]=O

Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics

Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Zeolite mordenite, ammonium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'