missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zeolite mordenite, ammonium

Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics

Brand:  Alfa Aesar™ 045877.22

Code : NEW

Additional Details : CAS Number : 1318-02-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15402517

  • £55.76 / 100g

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1318-02-1
162.043
HNPSIPDUKPIQMN-UHFFFAOYSA-N
9942228
O=[Al]O[Al]=O.O=[Si]=O
Al2O5Si
MFCD00132601
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
dioxosilane;oxo(oxoalumanyloxy)alumane

Specifications

Zeolite mordenite, ammonium
Al2O5Si
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
HNPSIPDUKPIQMN-UHFFFAOYSA-N
O=[Al]O[Al]=O.O=[Si]=O
162.043
Powder,S.A. 500 m
White
1318-02-1
MFCD00132601
Insoluble in water.
SiO2:Al2O3, 20:1 mole ratio
dioxosilane;oxo(oxoalumanyloxy)alumane
9942228
500m2/g
100g
Videos
SDS Documents
Certificates