Learn More
Zeolite beta, hydrogen, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)Zeolite β, hydrogen is used as a molecular sieve, filter, adsorbent, catalyst, drying agent, cation exchanger, dispersing agent and detergent builder. It serves as an acid catalyst and utilized as an alternative to the known methods for protection of alcohols as tetrahydropyranyl ethers and the deprotection of tetrahydropyranyl ethers. It is also used in the stereoselective Meerwein-Ponndorf-Verley reduction of ketones. Further, it is used as an efficient and recyclable catalyst for the tetrahydropyranylation of alcohols and phenols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
Specifications
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Zeolite beta, hydrogen |
RUO – Research Use Only