missing translation for 'onlineSavingsMsg'
Learn More

ZBTB40 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA550164

Additional Details : Weight : 0.01000kg

Product Code. 15654095

  • £329.00 / 100µL

Estimated Shipment: 16-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody detects endogenous levels of total ZBTB40 protein.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.
Specifications

Specifications

ZBTB40
Polyclonal
Unconjugated
ZBTB40
Liquid
ZBTB40
Synthesized peptide derived from C-terminal of human ZBTB40.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NUA8
KIAA0478
Rabbit
IgG
100 μL
-20°C
Primary
9923
Videos
SDS
Documents
Certificates