missing translation for 'onlineSavingsMsg'
Learn More

Zatebradine hydrochloride, Tocris Bioscience™

Blocks hyperpolarization-activated current (If)

Brand:  Tocris (Bio-Techne) 220250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13450794

  • £624.75 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Bradycardic agent that produces use-dependent inhibition of hyperpolarization-activated current If (HCN channel) in sinoatrial node cells (EC50 = 480 nM) and Purkinje fibres. Displays negative chronotropic activity in isolated guinea pig atria (EC50 of 13.4 μM).
Specifications

Chemical Identifiers

91940-87-3
493.041
zatebradine hydrochloride, unii-wjb7dyv16a, wjb7dyv16a, ul-fs49, dsstox_cid_25745, dsstox_rid_81090, dsstox_gsid_45745, zatebradine hcl, ul-fs 49
3-[3-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
C26H37ClN2O5
ZRNKXJHEQKMWCH-UHFFFAOYSA-N
3045335
CN(CCCN1CCC2=CC(=C(C=C2CC1=O)OC)OC)CCC3=CC(=C(C=C3)OC)OC.Cl

Specifications

50mg
91940-87-3
zatebradine hydrochloride, unii-wjb7dyv16a, wjb7dyv16a, ul-fs49, dsstox_cid_25745, dsstox_rid_81090, dsstox_gsid_45745, zatebradine hcl, ul-fs 49
CN(CCCN1CCC2=CC(=C(C=C2CC1=O)OC)OC)CCC3=CC(=C(C=C3)OC)OC.Cl
493.041
493.04
Zatebradine hydrochloride
C26H37ClN2O5
ZRNKXJHEQKMWCH-UHFFFAOYSA-N
3-[3-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
3045335
>98%
Videos
Documents