missing translation for 'onlineSavingsMsg'
Learn More

Zardaverine, Tocris Bioscience™

PDE3/4 inhibitor

Brand:  Tocris (Bio-Techne) 1046/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488373

  • £461.52 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Phosphodiesterase inhibitor, selective for PDE3 and 4 (IC50 values are 0.5 and 0.8μM respectively). Also available as part of the Phosphodiesterase Inhibitor Tocriset™ .
Specifications

Chemical Identifiers

101975-10-4
268.22
6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
CHEBI:46548
C12H10F2N2O3
HJMQDJPMQIHLPB-UHFFFAOYSA-N
5723

Specifications

Zardaverine
>98%
50mg
HJMQDJPMQIHLPB-UHFFFAOYSA-N
3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one
5723
268.22
101975-10-4
C12H10F2N2O3
6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F
268.22
CHEBI:46548
Videos