Learn More
ZAPA sulfate, Tocris Bioscience™
Agonist at 'low affinity' GABAA receptor. More potent than GABA/muscimol
Brand:  Tocris (Bio-Techne) 018010
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487453
  • £128.00 / 10mg
Estimated Shipment
01-03-2018
Add to basket
Description
Description
More potent than either GABA or muscimol as an agonist at low affinity GABAA receptors and is thus a useful ligand to investigate GABA receptors linked to benzodiazepine receptors. Also a GABAA-ρ receptor antagonist.
Specifications
Chemical Identifiers
ZAPA sulfate
C4H6N2O2S·H2SO4
244.24
(Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
Specifications
ZAPA sulfate
>98%
244.24
(Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
371962-01-5
C4H6N2O2S·H2SO4