missing translation for 'onlineSavingsMsg'
Learn More

Zamifenacin fumarate, Tocris Bioscience™

Selective M3 antagonist

Brand:  Tocris (Bio-Techne) 257950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461504

  • £633.57 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Selective M3 muscarinic receptor antagonist (pKi values are 8.52, 7.93, 7.90 and 7.78 for M3, M2, M1 and M4 receptors respectively). Displays higher affinity at ileal M3 receptors (pKi = 9.3) compared to oesophageal and tracheal M3 receptors (pKi values are 8.8 and 8.2 respectively). Inhibits gastrointestinal motility in vivo in the absence of cardiovascular effects.
Specifications

Chemical Identifiers

127308-98-9
531.605
zamifenacin fumarate
(3R)-3-benzhydryloxy-1-[2-(1,3-benzodioxol-5-yl)ethyl]piperidine;but-2-enedioic acid
C31H33NO7
PMHKTAIGYOVZEZ-GJFSDDNBSA-N
71433586
C1CC(CN(C1)CCC2=CC3=C(C=C2)OCO3)OC(C4=CC=CC=C4)C5=CC=CC=C5.C(=CC(=O)O)C(=O)O

Specifications

Zamifenacin fumarate
127308-98-9
zamifenacin fumarate
C1CC(CN(C1)CCC2=CC3=C(C=C2)OCO3)OC(C4=CC=CC=C4)C5=CC=CC=C5.C(=CC(=O)O)C(=O)O
531.605
531.6
50mg
C31H33NO7
PMHKTAIGYOVZEZ-GJFSDDNBSA-N
(3R)-3-benzhydryloxy-1-[2-(1,3-benzodioxol-5-yl)ethyl]piperidine;but-2-enedioic acid
71433586
>99%
Videos