missing translation for 'onlineSavingsMsg'
Learn More

Zacopride hydrochloride, Tocris Bioscience™

Highly potent 5-HT3 antagonist; also 5-HT4 agonist

Brand:  Tocris (Bio-Techne) 179550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13450014

  • £557.13 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent 5-HT3 receptor antagonist (Ki = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). Antiemetic and anxiolytic following systemic administration in vivo.
Specifications

Chemical Identifiers

101303-98-4
346.252
unii-bj3775635u component
4-amino-N-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-5-chloro-2-methoxybenzamide;hydrochloride
C15H21Cl2N3O2
ITXVOUSORXSTQH-BTQNPOSSSA-N
13272324
COC1=CC(=C(C=C1C(=O)NC2CN3CCC2CC3)Cl)N.Cl

Specifications

50mg
101303-98-4
unii-bj3775635u component
COC1=CC(=C(C=C1C(=O)NC2CN3CCC2CC3)Cl)N.Cl
346.252
346.26
Zacopride hydrochloride
C15H21Cl2N3O2
ITXVOUSORXSTQH-BTQNPOSSSA-N
4-amino-N-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-5-chloro-2-methoxybenzamide;hydrochloride
13272324
>99%
Videos
Documents