Learn More
Tocris Bioscience™ Z-Guggulsterone
Broad spectrum steroid receptor ligand. More active isomer of guggulsterone (Cat. No. 2013)
Brand:  Tocris Bioscience™ 357010
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463214
  • £149.85 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Broad spectrum steroid receptor ligand; mineralocorticoid, progesterone and glucocorticoid receptor antagonist (Ki values are 37, 224 and 252 nM respectively) and weak androgen receptor agonist (Ki = 315 nM). Induces apoptosis in prostate cancer cells and inhibits angiogenesis via suppression of the VEGF-VEGFR2-Akt signaling pathway. Exhibits antilipidemic activity via antagonism of the farnesoid X receptor (FXR) and displays antiseptic, antirheumatic and anti-inflammatory activity in vivo. More active isomer of guggulsterone (Cat. No. 2013).
Specifications
Specifications
Z-Guggulsterone
>98
10mg
Z-Guggulsterone
312.45