missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

Catalog Number p-8014494
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1g/L
Quantity 100 mL
Solution Contains HNO3
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'