missing translation for 'onlineSavingsMsg'
Learn More

Yttrium, plasma standard solution, Y 1000μg/mL, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 013855.AD

Code : NEW

Additional Details : Weight : 0.05000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 50mL

 View more versions of this product

Product Code. 11321429

  • £34.80 / 50mL


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

Yttrium
Liquid
MFCD00011468
7732-18-5
Y2O3 in 5% HNO3

Specifications

Yttrium
Liquid
Y2O3 in 5% HNO3
Nitric acid: 5.0%; Yttrium(III) oxide: 0.127%; Water: 94.873%
50mL
7732-18-5
Liquid
MFCD00011468
23 hPa (17mm Hg) at 20°C
Videos
SDS