missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Yttrium(III) neodecanoate, Y 12-16%

Brand:  Alfa Aesar™ 039566.09

Code : NEW

Additional Details : CAS Number : 68683-17-0 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11308388

  • £44.31 / 10g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

Yttrium(III) neodecanoate
68683-17-0
602.63
MFCD00070443

Specifications

Yttrium(III) neodecanoate
10g
Powder/Chunks
MFCD00070443
Y 12 to 16%
68683-17-0
602.63
Videos
SDS