missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) isopropoxide, 90+%, Thermo Scientific Chemicals

Catalog Number p-7049475
Click to view available options
Quantity:
1 g
5 g
2172-12-5
MFCD00015641
2172-12-5
MFCD00015641

Yttrium(III) isopropoxide is used in asymmetric catalysis applications. It acts as a catalyst for stereoselective conjugate addition reactions and ring-opening polymerizations. It serves as a precursor for the generation of reactive enolates by enantioselective protonation reactions and nanocomposites synthesis. Further, it is used in the preparation of flat hafnium(IV) oxide films. In addition to this, it is used to improve the thermal conductivity of polycrystalline aluminum nitride (AlN) ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 160°C
Boiling Point 200°C to 210°C (1 mmHg)(sublimation)
Molecular Formula C22H29KO4
Quantity 1 g
UN Number UN2925
Sensitivity Moisture sensitive
Solubility Information Soluble in hexane,toluene and terahydrofuran.
InChI Key JTZQCHFUGHIPDF-RYVBEKKQSA-M
SMILES CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC4(CCC(=O)[O-])O)C.[K+]
IUPAC Name potassium;3-[(10S,13R,17S)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
Molecular Weight (g/mol) 396.57
PubChem CID 133109093
Formula Weight 266.17
Percent Purity ≥90%
Physical Form Powder
Chemical Name or Material Yttrium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'