missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Yttrium(III) isopropoxide, 90+%

Brand:  Alfa Aesar™ 014477.06

Code : NEW

Additional Details : CAS Number : 2172-12-5 Weight : 0.00500kg Transport : UN number : 2925 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11312428

  • £193.30 / 5g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Yttrium(III) isopropoxide
266.17
UN2925
2172-12-5
MFCD00015641

Specifications

Yttrium(III) isopropoxide
200° to 210°C (1mmHg) (subl.)
5g
Powder
266.17
UN2925
Moisture Sensitive
160°C
2172-12-5
≥90%
MFCD00015641
Soluble in hexane, THF, toluene
Videos
SDS