missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) isopropoxide, 20-25% w/w in toluene, Thermo Scientific™

Catalog Number p-7049495
Click to view available options
Quantity:
∼1 g
∼25 g
5 g
This item is not returnable. View return policy
2172-12-5
C9H24O3Y
269.19
MFCD00015641
NREVZTYRXVBFAQ-UHFFFAOYSA-N
yttrium isopropoxide
85308566
propan-2-ol;yttrium
CC(C)O.CC(C)O.CC(C)O.[Y]
This item is not returnable. View return policy
2172-12-5
C9H24O3Y
269.19
MFCD00015641
NREVZTYRXVBFAQ-UHFFFAOYSA-N
yttrium isopropoxide
85308566
propan-2-ol;yttrium
CC(C)O.CC(C)O.CC(C)O.[Y]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 0.955 g/mL
Odor Aromatic
Linear Formula Y[OCH(CH3)2]3
Quantity ∼1 g
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information Soluble in hexane,toluene,and THF.
Formula Weight 266.17
Concentration or Composition (by Analyte or Components) 20 to 25% w/w in toluene
Physical Form Liquid
Chemical Name or Material Yttrium(III) isopropoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'