missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number 11326208
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
This item is not returnable. View return policy
12741-05-8
Cl3Y
195.26
MFCD00149941
PCMOZDDGXKIOLL-UHFFFAOYSA-K
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
57347377
yttrium(3+);trichloride;hydrate
[Cl-].[Cl-].[Cl-].[Y+3]
This item is not returnable. View return policy
12741-05-8
Cl3Y
195.26
MFCD00149941
PCMOZDDGXKIOLL-UHFFFAOYSA-K
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
57347377
yttrium(3+);trichloride;hydrate
[Cl-].[Cl-].[Cl-].[Y+3]

Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink to White
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula YCl3·xH2O
Merck Index 14,10107
Solubility Information Soluble in water,pyridine,ethanol.
Formula Weight 195.26 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 2.18g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'