missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Description
Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink to White |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Linear Formula | YCl3·xH2O |
Merck Index | 14,10107 |
Solubility Information | Soluble in water,pyridine,ethanol. |
Formula Weight | 195.26 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'