missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride hexahydrate, 99.999%, (trace metal basis), Thermo Scientific™

Catalog Number p-221579
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
10025-94-2
Cl3Y·6H2O
MFCD00149941
This item is not returnable. View return policy
10025-94-2
Cl3Y·6H2O
MFCD00149941

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 100.0°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 10 g
Assay Percent Range 100.00%
Linear Formula YCl3·6H2O
Merck Index 14, 10107
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 303.35
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass Bottle
Chemical Name or Material Yttrium(III) chloride hexahydrate, 99.999%
Show More Show Less
compliance-icons
Product Identifier
  • Yttrium(III) chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'