missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) bromide hydrate REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-7049186
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
MFCD00049710
This item is not returnable. View return policy
MFCD00049710

It is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 328.63 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) bromide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'