missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium fluoride, 99.99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
Description
For ultra trace analysis
Specifications
Quantity | 5 g |
Assay Percent Range | 99.99% |
Formula Weight | 145.9 |
Grade | Trace Metal Basis |
Packaging | Plastic bottle |
Chemical Name or Material | Yttrium fluoride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'