Learn More
Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Can be used as a sputtering target to prepare bubble memory devices.Yttrium aluminum oxide finds application in ceramic, glass and optics due to its thermal stability and insolubility. It is used in the preparation of sputtering bonded targets in chemical processes viz. chemical vapor deposition, atomic thermal and electron beam evaporation. It is also used in solar, water treatment and fuel cell. Further, it is used as coloring and adhesive agents in enamels manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1760°C |
Physical Form | Fused Lump |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 593.62 |
Percent Purity | 99.99% |
Refractive Index | 1.833 |
Density | 4.65 g/mL |
Assay | (metals basis) |
Chemical Name or Material | Yttrium aluminum oxide |
RUO – Research Use Only