missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals
108.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Rare earth element doped yttrium aluminum oxide is used in solid state lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1760°C |
Physical Form | Powder |
Quantity | 25 g |
Formula Weight | 593.62 |
Percent Purity | 99.9% |
Refractive Index | 1.833 |
Density | 4.65 g/mL |
Assay | (REO) |
Chemical Name or Material | Yttrium aluminum oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'